Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethylphenylammonium bromide | 16056-11-4 | 98.0% | 216.12 | 250 G
SDP

Catalog No. 5000453409
Encompass_Preferred

Trimethylphenylammonium bromide is a quaternary ammonium salt composed of a positively charged N,N,N-trimethylanilinium cation and a negatively charged bromide anion. It serves as a phase transfer catalyst in organic chemical reactions, aiding the transfer of reactants between immiscible phases. Additionally, it can be utilized as a reagent in the synthesis of various organic compounds and, in some applications, as a surfactant or corrosion inhibitor. It is a biochemical reagent suitable for life science research.

  • Appearance: Solid
  • Color: Light blue to blue
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years, or 4°C for 2 years. In solvent, -80°C for 6 months, or -20°C for 1 month
  • Solubility: In H2O: 50 mg/mL (231.35 mM; requires ultrasonic)

Catalog No. 50-004-53409 Supplier Medchemexpress LLC Supplier No. 250GHYW012450
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trimethylphenylammonium bromide is a quaternary ammonium salt composed of a positively charged N,N,N-trimethylanilinium cation and a negatively charged bromide anion. It serves as a phase transfer catalyst in organic chemical reactions, aiding the transfer of reactants between immiscible phases. Additionally, it can be utilized as a reagent in the synthesis of various organic compounds and, in some applications, as a surfactant or corrosion inhibitor. It is a biochemical reagent suitable for life science research.

  • Appearance: Solid
  • Color: Light blue to blue
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years, or 4°C for 2 years. In solvent, -80°C for 6 months, or -20°C for 1 month
  • Solubility: In H2O: 50 mg/mL (231.35 mM; requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.