Learn More
Strem, An Ascensus Company CAS# 512-56-1. 250g. Trimethylphosphate, min. 97%. MFCD00008348
SDP
Catalog No. 5090114233
Encompass
CAS# 512-56-1. 250g. Trimethylphosphate, min. 97%. MFCD00008348. Molecular Weight: 140.08. Molecular Formula: (CH3O)3P(O). Color/form: colorless liq. Strem# 93-1561. http://www.strem.com/catalog/v/93-1561/
Catalog No. 50-901-14233 Supplier Strem, An Ascensus Company Supplier No. 931561250G
CAS# 512-56-1. 250g. Trimethylphosphate, min. 97%. MFCD00008348. Molecular Weight: 140.08. Molecular Formula: (CH3O)3P(O). Color/form: colorless liq. Strem# 93-1561. http://www.strem.com/catalog/v/93-1561/