Learn More
Strem, An Ascensus Company Trimethylphosphine,min.98% 25g
SDP
Catalog No. 5090110446
Encompass
CAS# 594-09-2. 25g. Trimethylphosphine, min. 98%. MFCD00008510. Molecular Weight: 76.08. Molecular Formula: (CH3)3P. Color/form: colorless liq. Strem# 15-6500. http://www.strem.com/catalog/v/15-6500/
Catalog No. 50-901-10446 Supplier Strem, An Ascensus Company Supplier No. 15650025G
CAS# 594-09-2. 25g. Trimethylphosphine, min. 98%. MFCD00008510. Molecular Weight: 76.08. Molecular Formula: (CH3)3P. Color/form: colorless liq. Strem# 15-6500. http://www.strem.com/catalog/v/15-6500/