Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Trimethylphosphoranylidene)Acetonitrile Solution

Catalog No. 7529405ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-294-05ML 5 mL
75-294-010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 75-294-05ML Supplier Sigma Aldrich Supplier No. 7529405ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(Trimethylphosphoranylidene)acetonitrile is a reagent generally employed in the Wittig olefination of less reactive carboxylic acid compounds like esters, N-Boc lactams, lactones, and imides to give +a,+a-unsaturated nitriles. It is also used as an alternate reagent for Mitsonobu reaction by replacing DEAD/PPh3 for the alkylation of C-H and N-H bonds.If precipitate is observed, war mgently to resolubilize

Specifications

CAS 176325-83-0
Density 0.896 g/mL at 25°C
Molecular Formula C5H10NP
Linear Formula C5H10NP
MDL Number MFCD17013403
Quantity 5 mL
Synonym CMMP; Cyanomethylenetrimethylphosphorane solution
Molecular Weight (g/mol) 115.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.