Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylsilanol

Catalog No. 72598610ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-598-610ML 10 mL
72-598-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 72-598-610ML Supplier Sigma Aldrich Supplier No. 72598610ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylsilanol (TMS) can be used to prepare tris(trimethylsiloxy) aluminum (I) by reacting with triethylaluminum.

Specifications

CAS 1066-40-6
Molecular Formula C3H10OSi
Linear Formula (CH3)3SiOH
MDL Number MFCD02751657
Quantity 10 mL
Synonym TMS; TMSiOH; Hydroxytrimethylsilane
Molecular Weight (g/mol) 90.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.