Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific (Trimethylsilyl)acetylene, 1066-54-2, MFCD00008569, 25g

Catalog No. 500116696
Encompass_Preferred

Molecular Formula C5H10Si, Purity 98%, Molecular Weight 98.22, Boiling Point 53

Catalog No. 50-011-6696 Supplier Matrix Scientific Supplier No. 00434825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C5H10Si, Purity 98%, Molecular Weight 98.22, Boiling Point 53

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.