Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylsilyl cyanide

Catalog No. 7332850MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-328-50MLF 50 mL
73-328-250MLF 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 73-328-50MLF Supplier Sigma Aldrich Supplier No. 7332850MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylsilyl cyanide

Specifications

Boiling Point 114°C to 117°C (lit.)
Melting Point 8°C to 11°C (lit.)
CAS 7677-24-9
Quantity 50 mL
Molecular Formula C4H9NSi
Linear Formula (CH3)3 SiCN
MDL Number MFCD00001765
Synonym Cyanotrimethylsilane; TMSCN
Molecular Weight (g/mol) 99.21
Percent Purity ≥95% (GC)
Refractive Index n20/D 1.392 (literature)
Density 0.793 g/mL (at 20°C (literature))
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.