Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylsilyl cyclopentadiene, mixture of isomers

Catalog No. 25788510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-788-510G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-788-510G Supplier Sigma Aldrich Supplier No. 25788510G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylsilyl cyclopentadiene, mixture of isomers

Specifications

CAS 3559-74-8
Density 0.833 g/mL (at 25°C (literature))
Boiling Point 138°C to 140°C (lit.)
Molecular Formula C8H14Si
Refractive Index n20/D 1.471 (literature)
Linear Formula C8H14Si
MDL Number MFCD00010666
Quantity 100 mg
Synonym NSC 176274
Molecular Weight (g/mol) 138.28
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.