Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Trimethylsilyl)isocyanate

Catalog No. 25264625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-264-625G 25 g
25-264-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-264-625G Supplier Sigma Aldrich Supplier No. 25264625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trimethylsilyl isocyanate was used in:
  • Postsynthetic modification of isoreticular metal-organic framework-3
  • Synthesis of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones
  • Conversion of isocyanates to carbodiimides with cyclopentadienyl fe(co)2 or -mn(co)3 catalysis
  • Carbamoylation of aromatic hydrocarbons and alcohols

Specifications

CAS 1118-02-1
Density 0.851 g/mL (at 25°C (literature))
Boiling Point 90°C to 92°C (lit.)
Molecular Formula C4H9NOSi
Refractive Index n20/D 1.396 (literature)
Linear Formula (CH3)3 SiNCO
MDL Number MFCD00001993
Quantity 25 g
Synonym Isocyanatotrimethylsilane
Molecular Weight (g/mol) 115.21
Percent Purity 85%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.