Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Trimethylsilyl methylam | 25ml | 18166-02-4 | MFCD00042931

Supplier:  Sigma Organic Chemistry 9276825ML

Encompass_Preferred

Trimethylsilyl methylam , 25ml

About this item:

CAS #: 18166-02-4
MDL #: MFCD00042931
Purity: 0.98
Molecular Weight: 103.24
UNSPSC Code: 12352100

Catalog No. 92-768-25ML


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.