Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Trimethylsilyl)methylamine

Catalog No. 502907962
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7962 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-7962 Supplier Sigma Aldrich Supplier No. 9276825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Synthesis of trimethylsilyl-diazomethane; 1,3-Cycloaddition of its imine; preparation of azomethine ylides from ketones
TRUSTED_SUSTAINABILITY

Specifications

CAS 18166-02-4
Density 0.771 g/mL (at 20°C (literature))
Boiling Point 92°C to 94°C (lit.)
Molecular Formula C4H13NSi
Refractive Index n20/D 1.416
Linear Formula C4H13NSi
MDL Number MFCD00042931
Quantity 25 mL
Synonym (Aminomethyl)trimethylsilane
Molecular Weight (g/mol) 103.24
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.