Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Trimethylsilyl)Methyllithium Solution

Catalog No. 29705450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
4 x 25 mL
600 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-705-450ML 50 mL
29-705-4600ML 600 mL
29-705-425ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 29-705-450ML Supplier Sigma Aldrich Supplier No. 29705450ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Initiates polymerization and copolymerization of cyclosiloxanes. Alkylates metal complexes.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1822-00-0
Density 0.65 g/mL at 25°C
Boiling Point 35°C to 36°C
Molecular Formula C4H11LiSi
Linear Formula (CH3)3SiCH2Li
MDL Number MFCD00010747
Quantity 50 mL
Molecular Weight (g/mol) 94.16 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.