Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Trimethylsilyl)Methylmagnesium Chloride Solution

Catalog No. 503081440 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1440 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 50-308-1440 Supplier Sigma Aldrich Supplier No. 256021100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used in a variety of nickel catalyzed reactions including alkenation of dithioacetals, cross-coupling with aryl triflates and carbamates, and cross-coupling with vinyl selenides.

Specifications

CAS 1588-83-6
Melting Point 280°C
Molecular Formula C7H6N2O4
Linear Formula H2NC6H3(NO2)CO2H
MDL Number MFCD00017009
Quantity 25 g
Molecular Weight (g/mol) 182.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.