Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylsilyl Trifluoromethanesulfonate

Catalog No. 9174150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-741-50ML 50 mL
91-741-10ML 10 mL
91-741-250ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 91-741-50ML Supplier Sigma Aldrich Supplier No. 9174150ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylsilyl trifluoromethanesulfonate may be used to catalyze:Allylation of acetals to form homoallyl ethers.Synthesis of 1,2-trans-glycosides.Conversion of alcohols to esters.Aminomethylation of silyl enol ethers with aminomethyl alkyl ethers.Glycosidation of (+)-4-demethoxyanthracyclinones.Efficient silylating agent and strong Lewis acid catalyst ; Review

Specifications

CAS 27607-77-8
Density 1.228 g/mL at 25°C
Boiling Point 77°C80 mmHg, lit.)
Molecular Formula C4H9F3O3SSi
Refractive Index n20/D 1.36
Linear Formula CF3SO3Si(CH3)3
MDL Number MFCD00000406
Quantity 50 mL
Synonym TMS triflate; TMSOTf; Trifluoromethanesulfonic acid trimethylsilylester
Molecular Weight (g/mol) 222.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.