Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylsilyl Trifluoromethanesulfonate

Catalog No. 225649250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-564-9250G 250 g
22-564-950G 50 g
22-564-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 22-564-9250G Supplier Sigma Aldrich Supplier No. 225649250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylsilyl trifluoromethanesulfonate has been used in combination with boron trifluoride etherate for the copper-catalyzed asymmetric allylic alkylation (AAA) of allyl bromides, chlorides, and ethers with organolithium reagents in the presence of a chiral ligand.It can be used:As a silylating agent for the synthesis of trimethylsilyl-enol ethers from esters of +a-diazoacetoacetic acid.To activate benzyl and allyl ethers for the alkylation of sulfides.To facilitate the conversion of Diels-Alder adducts of Danishefsky's diene to cyclohexenones without the formation of methoxy ketone by-product.To prepare difluoroboron triflate etherate, a powerful Lewis acid especially in acetonitrile solvent. As a reagent in a Dieckmann-like cyclization of ester-imides and diesters.It may also be used to catalyze:Acylation of alcohols with acid anhydrides.Reductive coupling of carbonyl compounds with trialkylsilanes to form symmetrical ethers.Glycosidation of 4-demethoxydaunomycinones with 1-O-acyl-L-daunosamine derivatives.

Specifications

CAS 27607-77-8
Density 1.228 g/mL at 25°C
Boiling Point 77°C80 mmHg, lit.)
Molecular Formula C4H9F3O3SSi
Refractive Index n20/D 1.36
Linear Formula CF3SO3Si(CH3)3
MDL Number MFCD00000406
Quantity 250 g
Synonym TMS triflate; TMSOTf; Trifluoromethanesulfonic acid trimethylsilylester
Molecular Weight (g/mol) 222.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.