Learn More
Strem, An Ascensus Company CAS# 1066-54-2. 5g. Trimethylsilylacetylene, min. 97%. MFCD00008569
SDP
Catalog No. 5090109633
Encompass
CAS# 1066-54-2. 5g. Trimethylsilylacetylene, min. 97%. MFCD00008569. Molecular Weight: 98.22. Molecular Formula: (CH3)3SiC≡CH. Color/form: colorless liq. Strem# 14-8200. http://www.strem.com/catalog/v/14-8200/
Catalog No. 50-901-09633 Supplier Strem, An Ascensus Company Supplier No. 1482005G
CAS# 1066-54-2. 5g. Trimethylsilylacetylene, min. 97%. MFCD00008569. Molecular Weight: 98.22. Molecular Formula: (CH3)3SiC≡CH. Color/form: colorless liq. Strem# 14-8200. http://www.strem.com/catalog/v/14-8200/