Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylstearylammonium Chloride 98.0+%, TCI America™
SDP

Catalog No. S0087100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0087100G 100 g
S008725G 25 g
S0087500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0087100G Supplier TCI America Supplier No. S0087100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-03-8
Molecular Formula C21H46ClN
Molecular Weight (g/mol) 348.06
MDL Number MFCD00050188
InChI Key VBIIFPGSPJYLRR-UHFFFAOYSA-M
Synonym n,n,n-trimethyloctadecan-1-aminium chloride, stac, octadecyltrimethylammonium chloride, cation ab, stearyltrimethylammonium chloride, steartrimonium chloride, trimethyloctadecylammonium chloride, 1-octadecanaminium, n,n,n-trimethyl-, chloride, nissan cation ab, aliquat 7
PubChem CID 8155
ChEBI CHEBI:81710
IUPAC Name trimethyl(octadecyl)azanium chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Quantity 100 g
Formula Weight 348.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylstearylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber BT1750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.