Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsulfonium bromide, 98%

Catalog No. p-7027997
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2509814 25 g
AAB2509822 100 g
AAB2509836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2509814 Supplier Thermo Scientific Chemicals Supplier No. B2509814
Only null left
Add to Cart

CAS: 3084-53-5 | C3H9BrS | 157.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3084-53-5
Molecular Formula C3H9BrS
Molecular Weight (g/mol) 157.069
MDL Number MFCD00051367
InChI Key GOTIICCWNAPLMN-UHFFFAOYSA-M
Synonym trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f
PubChem CID 2735154
IUPAC Name trimethylsulfanium;bromide
SMILES C[S+](C)C.[Br-]

Specifications

Melting Point 203°C (sublimation)
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 157.07
Percent Purity 98%
Chemical Name or Material Trimethylsulfonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.