Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyltin chloride, 99%

Catalog No. AC163990100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163990100 10 g Glass bottle
AC163990500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163990100 Supplier Thermo Scientific Chemicals Supplier No. 163990100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-45-1
Molecular Formula C3H11ClSn
Molecular Weight (g/mol) 201.28
MDL Number MFCD00000520
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
PubChem CID 14016
IUPAC Name chloro(trimethyl)stannane
SMILES Cl.C[SnH](C)C

Specifications

Melting Point 37.0°C to 39.0°C
Color White
Boiling Point 148.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SnCl
Quantity 10 g
Fieser 17,388; 03,19; 04,27; 05,26; 06,41; 07,18; 13,239
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,organic solvents
Formula Weight 199.25
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Trimethyltin chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
EINECSNumber 213-917-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.