Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyltin Chloride Solution

Catalog No. 37518725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-518-725ML 4 x 25 mL
37-518-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 37-518-725ML Supplier Sigma Aldrich Supplier No. 3751874X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyltin chloride solution (1.0 M in hexanes) can be used as a reagent in the synthesis of conjugated polymers based on isoindigo as acceptor and syn- or anti-benzo[1,2-b:5,4-b'sa]difuran (BDF) as a donor by Stille cross-coupling reaction. It can further be used for the investigation of optical, electrochemical and photovoltaic properties.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1066-45-1
Density 0.797 g/mL at 25°C
Molecular Formula C3H9ClSn
Linear Formula (CH3)3SnCl
Quantity 4 x 25 mL
MDL Number MFCD00000520
Molecular Weight (g/mol) 199.27 g/mol
Biological Activity Central Nervous System, Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.