Learn More
Cayman Chemical Trimidox hydrochloride
SDP
Catalog No. 502888894
Encompass_Preferred
A specific ribonucleotide reductase inhibitor; inhibits growth of human promyelocytic leukemia HL-60 cells (IC50 = 35 µM)
Catalog No. 50-288-8894 Supplier Cayman Chemical Supplier No. 1001112450 MG
A specific ribonucleotide reductase inhibitor; inhibits growth of human promyelocytic leukemia HL-60 cells (IC50 = 35 µM)