Learn More
Cayman Chemical Trimidox (hydrochloride), Purity: ≥98%, CAS: 95933-75-8, 10 mg
SDP
Catalog No. 502887284
Encompass_Preferred
A specific ribonucleotide reductase inhibitor; inhibits growth of human promyelocytic leukemia HL-60 cells (IC50 = 35 µM)
Catalog No. 50-288-7284 Supplier Cayman Chemical Supplier No. 1001112410 MG
A specific ribonucleotide reductase inhibitor; inhibits growth of human promyelocytic leukemia HL-60 cells (IC50 = 35 µM)