Learn More
TARGETMOL CHEMICALS INC Trimipramine maleate 500MG
SDP
Catalog No. 502880538
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Trimipramine maleate (Surmontil maleate) appears to inhibit serotonin transport and norepinephrine uptake by nerve terminals. This increases available norepinephrine or serotonin and prolongs its action. Trimipramine Maleate is the maleate salt form of trimipramine, a tricyclic secondary amine of the dibenzazepine class, with an antidepressant property. Purity 98.02%
Catalog No. 50-288-0538 Supplier TARGETMOL CHEMICALS INC Supplier No. T1045500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Trimipramine maleate (Surmontil maleate) appears to inhibit serotonin transport and norepinephrine uptake by nerve terminals. This increases available norepinephrine or serotonin and prolongs its action. Trimipramine Maleate is the maleate salt form of trimipramine, a tricyclic secondary amine of the dibenzazepine class, with an antidepressant property. Purity 98.02%