Learn More
MSC Carlisle Lid, Swing Top, Rectangle, 15 and 23 Gallon, ABS plastic, Black
Catalog No. 502839136
Encompass
This rectangular swing top lid is designed for hands-free waste disposal. Its slim profile allows it to fit into narrow spaces, making it ideal for various environments. The lid is made from durable ABS plastic and is compatible with both 15- and 23-gallon waste containers.
  • Slim shape allows it to fit into narrow spaces along walls and near work benches.
  • Hinged swing door enables hands-free waste disposal.
  • Compatible with both 15- and 23-gallon waste containers.
  • Made from recyclable plastic, type "7".
Catalog No. 50-283-9136 Supplier MSC Supplier No. 73916090
This rectangular swing top lid is designed for hands-free waste disposal. Its slim profile allows it to fit into narrow spaces, making it ideal for various environments. The lid is made from durable ABS plastic and is compatible with both 15- and 23-gallon waste containers.
  • Slim shape allows it to fit into narrow spaces along walls and near work benches.
  • Hinged swing door enables hands-free waste disposal.
  • Compatible with both 15- and 23-gallon waste containers.
  • Made from recyclable plastic, type "7".