Learn More
Belair Instrument Company Inc Avantik GR2094-A Trimming Knife, Stainless Steel Blades, 130 mm Length
SDP
Catalog No. NC1282892
Encompass_Preferred
The Avantik trimming knife is a precision-engineered tool featuring stainless steel blades, ideal for exact trimming and grossing in pathology labs. With a length of 130 mm and packaged in a secure dispenser, it ensures safety and maintains sharpness. These durable blades provide precise cutting for various tissues, differentiating themselves through compatibility with standard handles like Feather and Tissue-Tek, making them a reliable choice for professionals.
Catalog No. NC1282892 Supplier Belair Instrument Company Inc Supplier No. GR2094A
The Avantik trimming knife is a precision-engineered tool featuring stainless steel blades, ideal for exact trimming and grossing in pathology labs. With a length of 130 mm and packaged in a secure dispenser, it ensures safety and maintains sharpness. These durable blades provide precise cutting for various tissues, differentiating themselves through compatibility with standard handles like Feather and Tissue-Tek, making them a reliable choice for professionals.