Learn More
Andwin Scientific Tissue-Tek Accu-Edge Trimming Knife
SDP
Catalog No. 50727506
Encompass_Preferred
The Tissue-Tek Accu-Edge Trimming Knife Handles feature very sharp points and a lightweight design. This product is ideal for surgical grossing and autopsy procedures. The sharp edge pemits a broad ways of sectioning and trimming tissue samples. This product is described as long and straight. It is sold individually.
Catalog No. 50-727-506 Supplier Andwin Scientific Supplier No. 6256004
The Tissue-Tek Accu-Edge Trimming Knife Handles feature very sharp points and a lightweight design. This product is ideal for surgical grossing and autopsy procedures. The sharp edge pemits a broad ways of sectioning and trimming tissue samples. This product is described as long and straight. It is sold individually.