Learn More
Andwin Scientific TRIMMING KNIFE SHORT STRAIGHT
SDP
Catalog No. 50727505
Encompass
Lightweight Sakura Finetek Short-Straight trimming knives are designed for surgical grossing and autopsy procedures. Its highly sharp edges permit a variety of section and trim sampling. Disposable blades can be inserted easily into handle.
This item is not returnable. View return policy
Catalog No. 50-727-505 Supplier Andwin Scientific Supplier No. 6256001
Lightweight Sakura Finetek Short-Straight trimming knives are designed for surgical grossing and autopsy procedures. Its highly sharp edges permit a variety of section and trim sampling. Disposable blades can be inserted easily into handle.