Learn More
Electron Microscopy Sciences Trimming Knives, 50/PK
SDP
Supplier:  Electron Microscopy Sciences 6256201
Encompass
The highest quality, longest lasting trimming knives are made from stainless steel. The blades fit the Feather handle and the Tissue-Tek handle and are ideal for precise and sharp trimming and grossing. Available in 130 mm and 260 mm lengths. OAL: 130 mm.
Catalog No. 50-209-0708