Learn More
Electron Microscopy Sciences Trimming Knives, 50/PK
SDP
Catalog No. 502090709
Encompass
The highest quality, longest lasting trimming knives are made from stainless steel. The blades fit the Feather handle and the Tissue-Tek handle and are ideal for precise and sharp trimming and grossing. Available in 130 mm and 260 mm lengths. OAL: 260 mm.
Catalog No. 50-209-0709 Supplier Electron Microscopy Sciences Supplier No. 6256202
The highest quality, longest lasting trimming knives are made from stainless steel. The blades fit the Feather handle and the Tissue-Tek handle and are ideal for precise and sharp trimming and grossing. Available in 130 mm and 260 mm lengths. OAL: 260 mm.