Learn More
Surgical Design™ Trimming Microtome Blades and Handles
Catalog No. 21124907 Shop All Surgical Design Products
Change view
Click to view available options
Profile:
Hand
High
Low
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile
21-124-907 Hand
21-124-905 Low
21-124-906 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-124-907 Supplier Surgical Design™ Supplier No. HTPT003
Add to Cart
Add to Cart
Trimming microtome blades deliver clean, consistent tissue sectioning, with compatible handles for secure, stable blade control.
  • Available in Low Profile and High Profile
  • Ultra-sharp edges for smooth, uniform tissue trimming
  • Designed for paraffin blocks and routine histology section preparation
  • Compatible with most standard microtome blade holders
  • Reduced chatter and drag for cleaner ribbon formation
  • Individually wrapped for safety and contamination control
  • Stainless steel construction with long-lasting sharpness
  • Ideal for histology, pathology labs, medical research, forensic analysis, and autopsy/postmortem tissue sectioning
  • Provides controlled, ergonomic grip for safe blade manipulation
  • Compatible with low-profile and high-profile trimming blades
  • Rigid stainless-steel build for stability and durability
  • Textured grip surface enhances handling during precision trimming
  • Autoclavable, corrosion-resistant, and reusable
  • Suitable for specimen block trimming in histology, pathology, and postmortem tissue preparation
Specifications
Product Type Handle
Profile Hand
For Use With (Equipment) High and Low Profile Microtome
Disposable Single-use