Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma Pre-set crystals, 250 g
Catalog No. NC1537503
Encompass
BioPerformance Certified, pH 7.4, average Mw 151.6, Synonym: Tris crystals. The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.
Catalog No. NC1537503 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T7693250G
BioPerformance Certified, pH 7.4, average Mw 151.6, Synonym: Tris crystals. The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.