Learn More
Enzo Life Sciences Trinactin (1mg). CAS: 7561-71-9
SDP
Supplier:  Enzo Life Sciences ALX380238M001
Encompass_Preferred
Cation ionophore with high selectivity for ammonium and potassium. Immunosuppressive agent. Alternative name: Antibiotic AKD 1D, Antibiotic S 3466B. Purity: ≥95% (HPLC). Solubility: Soluble in ethanol, methanol, DMF or DMSO. Limited water solubility. Long Term Storage: -20°C.
Catalog No. 50-200-8986