Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trinity Biotech Buffers for Premier Hb9210 Analyzers

Catalog No. 22140172
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Premier™ Buffer A
Premier™ Buffer B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-140-172 Premier™ Buffer A
22-140-173 Premier™ Buffer B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-140-172 Supplier Trinity Biotech Supplier No. 01030095
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the quantitative measurement of hemoglobin A1c in human capillary and venous whole blood

  • Performance validated to assure accuracy and precision
  • Ready for use
  • 940mL

Specifications

Content And Storage 15°C to 28°C
Type Premier™ Buffer A
For Use With (Application) Quantitative measurement of hemoglobin A1c, in-vitro diagnostic use only
Quantity 940 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.