Learn More
Trinity Biotech Glycated Hemoglobin (HbA1c) Calibrator
Catalog No. 22140149
Click to view available options
Quantity:
Each
Catalog No. Quantity
22-140-149 Each
1 options
This item is not returnable. View return policy
Catalog No. 22-140-149 Supplier Trinity Biotech Supplier No. 01040018
This item is not returnable. View return policy
  • For use with Ultra2, PDQ and PDQ Plus systems
  • For quantitative measurement of the percentage of total glycated hemoglobins in whole blood
  • For in vitro diagnostic usage only
  • 2x2 Vials
Specifications
Type Glycated Hemoglobin Calibrator Kit
For Use With (Application) Calibration of quantitative HbA1c affinity assays, in-vitro diagnostic use only
Includes 1 x 400μL Level 1, 1 x 400μL level 2 calibrators
Quantity Each