Learn More
Trinity Biotech Glycated Hemoglobin (HbA1c) Control
Catalog No. 22140148
Click to view available options
Quantity:
Each
Catalog No. Quantity
22-140-148 Each
1 options
This item is not returnable. View return policy
Catalog No. 22-140-148 Supplier Trinity Biotech Supplier No. 01040015
This item is not returnable. View return policy
Intended for quantitative measurement of glycated haemoglobin in patient's blood
  • Used with Trinity Biotech ultra2, PDQ and PDQ Plus systems
  • Useful mean of diagnosing diabetes and subsequently assessing glycaemic control in diabetics
  • For in-vitro diagnostic use only
  • 2 x 2 Vials
Specifications
Type Glycated Hemoglobin (HbA1c) Control Kit
For Use With (Application) Control of quantitative HbA1c affinity assays, in-vitro diagnostic use only
Includes 1 x 400μL Level 1, 1 x 400μL level 2 control
Quantity Each