Learn More
Trinity Biotech Premier Wash Reagent
Catalog No. 22140175
Click to view available options
Quantity:
940 mL
Catalog No. Quantity
22-140-175 940 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-140-175 Supplier Trinity Biotech Supplier No. 01030098
This item is not returnable. View return policy
Designed for use with the Trinity Biotech Premier Hb9210 system
  • Glycated haemoglobin
  • For the quantitative measurement of the percentage of total glycated hemoglobins in whole blood
  • For in vitro diagnostic use only
Certifications
CE marked, Canadian Lisenced
Compliance
FDA compliance
Specifications
Type Wash Reagent
For Use With (Application) Quantitative measurement of the percentage of total glycated hemoglobins in whole Blood, For in-vitro diagnostic use only
Quantity 940 mL