Learn More
MSC Trinity Brand Industries Feeler Stock Roll, 0.0005 in x 1/2 in, 10 ft, Stainless Steel
Catalog No. 5000533520
Encompass
This Trinity Brand Industries Feeler Stock Roll is designed for precision measurements and adjustments in various applications. Made from durable stainless steel, it offers accurate thickness and consistency for reliable performance.
  • Constructed from stainless steel for durability and corrosion resistance.
  • Provides precise thickness of 0.0005 inch for accurate gaging.
  • Features a width of 1/2 "
  • Supplied as a 10 feet long roll for versatile applications.
Catalog No. 50-005-33520 Supplier MSC Supplier No. 01378819
This Trinity Brand Industries Feeler Stock Roll is designed for precision measurements and adjustments in various applications. Made from durable stainless steel, it offers accurate thickness and consistency for reliable performance.
  • Constructed from stainless steel for durability and corrosion resistance.
  • Provides precise thickness of 0.0005 inch for accurate gaging.
  • Features a width of 1/2 "
  • Supplied as a 10 feet long roll for versatile applications.