Learn More
Motic Swift Line Trinocular Cordless LED Microscope with 100X
Catalog No. S001200 Shop All Motic Swift Line Products
Change view
Click to view available options
Eyepiece:
10X/18 mm
Catalog No. Eyepiece
S001200 10X/18 mm
1 options
Catalog No. S001200 Supplier Motic Swift Line Supplier No. M3802CT4
Add to Cart
Add to Cart
Combining a new design with built-in handle to encourage proper handling, and semi-plan objectives, this microscope fits the needs of taxing clinical and student environments.
The Swift M3800 series continues the Swift tradition for value and quality with a brand new exciting look! Packed with standard research features, this series is ideal for advanced high school, biology, college, veterinarian, medical or technical applications.
  • OPTICAL SYSTEM:
  • Widefield 10x /18 mm eyepieces.
  • Trinocular viewing head with 45° inclined eyepieces, with diopter and interpupillary adjustment 54 mm to 76 mm.
  • Infinity plan 4x (0.10 N.A.), 10x (0.25 N.A.), 40xR (0.65 N.A.), and 100xR (1.25 N.A.) objectives. All are parfocalled, parcentered and color-coded.
  • Quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives. Mounted in reverse position to facilitate ease in changing slides.
  • ILLUMINATION:
  • LED light rated up to 50,000 hours of life, illumination equal to a 20 W tungsten bulb.
  • Batteries allow up to 500 recharges, for a total of 20,000 to 25,000 hours use from a single set of 3 AA rechargeable nickel hydride batteries. Battery charger included with microscope, automatically shuts off when batteries are fully charged, no danger of overcharging batteries and shortening battery life. No expensive recharger station needed/ Plug directly into wall outlet, or use ordinary, inexpensive power strip for recharging multiple microscopes. Indicator light changes from red to green when fully charged.
  • Single 8 hour charge allows up to 50 hours of use.
  • ELECTRICAL:
  • 110 V to 220 V switching power supply, unit shipped with American plug.
  • FOCUSING:
  • Coaxial coarse and fine focusing controls, coarse has tension adjustment, fine is graduated.
  • STAGE:
  • Large 5-3/4 in. x 4-3/4 in. (143 mm x 120 mm) stage; built-in, low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls.
  • Built-in 1.25 Abbe condenser has iris diaphragm.
  • FRAME:
  • Base dimensions: 9-1/4 in. x 6 in. (235 mm x 151 mm)
  • Height: 14-1/4 in. (362 mm). Net wt. 11 lb. (24.3 kg).
Trinocular Cordless LED Microscope, LED Microscope, Motic microscope, microscope
Specifications
Eyepiece 10X/18 mm
Head Style Trinocular
Lighting Type LED
Objective Types Infinity Corrected Plan
Product Line Swift Line
Product Type Trinocular Biological Microscope
Voltage 110/220 V