Learn More
Microscope World Fein Optic Trinocular Lab Microscope
SDP
Supplier:  Microscope World RB20T
Encompass_Preferred
The Fein Optic RB20 high power laboratory biological microscope offers high quality optics at an affordable price point. Objective Lenses- Infinity Corrected DIN Plan Achromat 4x, 10x, 40xR, 100xR (oil immersion) objectives. All microscope objectives are parfocalled, parcentered and color-coded. Quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives, features textured rubber knurling for easy grip. Mounted in reverse position to facilitate ease in changing slides.Eyepieces- Widefield High Eyepoint Focusing 10x eyepieces / FN 20mm. Eyepiece can hold a 20mm diameter reticle if the reticle retaining ring is added.Viewing Head- Trinocular Siedentopf viewing head with 30° inclined binocular eyepiece with interpupillary adjustment 47mm to 75mm. Vertical trinocular tube for camera integration.Total Magnification- 40x, 100x, 400x, 1000x.Size & Weight- 9in W x 15.5in D x 14in H, 17 lbs.Warranty- 5 Year warranty, excluding bulb, cord and fuse.
Catalog No. NC2965676