Learn More
ACCU-SCOPE Trinocular Microscope with HDS Camera and Screen
SDP
Catalog No. 01189347 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Eyepiece:
WF 10X/22 mm
Catalog No. Eyepiece
01-189-347 WF 10X/22 mm
1 options
Catalog No. 01-189-347 Supplier ACCU-SCOPE Supplier No. EXI310PH40MSHDSLT
Add to Cart
Add to Cart
Request a Quote
Provides reliable performance for tissue culture and clinical use with clear, high-contrast imaging of live cells.
  • Trinocular viewing head, inclined 45°, light distribution binocular 100% or Binocular/trinocular tube 80%/20%
  • WF10X/22 mm eyepieces with roll down eyeguards
  • Side facing quintuple nosepiece
  • Infinity Corrected Plan Achromat objectives (4X, 10X, 20X, 40X) and a mechanical stage
  • Excelis 1080P HD Camera with integrated 1080p HDMI display screen, 13.3 in., 60FPS USB 2.0 Output, SD Card 1/2.8 in. CMOS Sensor, USB 2.0, SD Card, Bracket
  • Mechanical stage with universal holder
  • 40X phase contrast objective
  • ELWD condenser, N.A. 0.3, Working Distance = 72 mm
  • Phase slider with 4X/10X, 20X/40X phase annulli, and open position
  • Phase centering telescope
  • Variable 5 Watt LED illuminator
  • Dust cover, power cord, Universal power supply 100 - 240 V
Specifications
Voltage 110/240 V
Magnification Power 4X, 10X, 20X, 40X
Objective Types Infinity Corrected Plan Achromatic, Phase Contrast
Eyepiece WF 10X/22 mm
Lighting Type LED
Head Style Trinocular, 45° Inclined
Camera Type Excelis HDS
Display Type Digital
Model ACCU-SCOPE
Product Type Inverted Phase-contrast Microscope