Learn More
ACCU-SCOPE Trinocular Microscope with Infinity-Corrected Plan Objectives and Excelis HDS Lite Camera System
SDP
Catalog No. 01189336 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Magnification Power:
4X, 10X, 40XR, 100XR
Catalog No. Magnification Power
01-189-336 4X, 10X, 40XR, 100XR
1 options
Catalog No. 01-189-336 Supplier ACCU-SCOPE Supplier No. EXC350HDSLT
Add to Cart
Add to Cart
Offers high optical performance, versatility, and durability for clinical, veterinary, and educational laboratories.
  • Designed for high optical performance, versatility, and long-term durability, making it ideal for clinical, veterinary, and educational laboratory environments.
  • Trinocular viewing head inclined at 30° and fully rotatable 360° enables ergonomic viewing and simultaneous digital imaging.
  • WF10X/20 mm focusing eyepieces with eyeguards provide enhanced clarity and user comfort during extended observation.
  • Reversed quintuple nosepiece allows smooth and efficient switching between objectives.
  • Infinity-corrected PLAN objectives (4X, 10X, 40XR, and 100XR oil) ensure sharp, high-resolution images with excellent flatness across the field.
  • Mechanical stage with low-position coaxial controls and right-hand operation supports precise and user-friendly slide manipulation.
  • Integrated Excelis HDS Lite camera system with monitor and 0.5X adapter delivers bright images and supports efficient digital workflows.
  • N.A.1.25 Abbe condenser with iris diaphragm provides optimal illumination control for enhanced contrast.
  • Coaxial coarse and fine focusing controls allow smooth, accurate focusing adjustments.
  • 3W variable LED illumination with field iris ensures consistent lighting, complemented by a protective dust cover.
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Trinocular, 30° Inclined, 360° Rotating
Lighting Type LED
Magnification Power 4X, 10X, 40XR, 100XR
Model ACCU-SCOPE
Objective Types Infinity Corrected Plan
Product Type Trinocular Microscope
Voltage 110/240 V