Learn More
ACCU-SCOPE™ Trinocular Microscope with Infinity Plan Achromat 4x, 10x and 40xR Objectives
SDP
Catalog No. 01000947 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Trinocular
Catalog No. Head Style
01-000-947 Trinocular
1 options
Catalog No. 01-000-947 Supplier ACCU-SCOPE™ Supplier No. 3001LED40
The 3001-LED-40 features a Trinocular microscope with Infinity Plan Achromat 4x, 10x, and 40xR objectives.
  • Trinocular viewing head (50/50 light split)
  • Optional C-mount or photo tube adapters sold separately
  • HWF Plan 10x/20mm focusing eyepieces (paired - one w/pointer), with eyeguards
  • Reversed quadruple nosepiece
  • Infinity plan 4x, 10x and 40xR
  • Mechanical stage, 140 X 132 mm
  • Abbe condenser, N.A. 1.25 with iris diaphragm (with slot for sliders)
  • Coaxial coarse and fine focusing controls
  • 3 watt LED variable illuminator
  • Dust cover, power cord, immersion oil and instruction manual
Specifications
Eyepiece HWF 10X/20 mm
Head Style Trinocular
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types Infinity Plan Achromatic
Product Type Trinocular LED Microscope
Voltage 110/240 V