Learn More
ACCU-SCOPE Trinocular Microscope with Plan 4X, 10X, and 40XR Objectives
SDP
Catalog No. 01189324 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Magnification Power:
4X, 10X, 40XR
Catalog No. Magnification Power
01-189-324 4X, 10X, 40XR
1 options
Catalog No. 01-189-324 Supplier ACCU-SCOPE Supplier No. EXC35040
Add to Cart
Add to Cart
Delivers high optical performance, versatility, and durability for clinical, veterinary, and educational labs.
  • Trinocular viewing head inclined 30° rotatable 360°
  • WF10X20 mm focusing eyepieces with eyeguards
  • Reversed quintuple nosepiece
  • Infinity corrected PLAN 4X 10X 40X R
  • Mechanical stage with low position coaxial controls right hand control
  • NA 1.25 Abbe condenser with iris diaphragm
  • Coaxial coarse and fine focusing controls
  • 3 watt variable LED with field iris and dust cover
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Trinocular, 30° Inclined, 360° Rotating
Lighting Type LED
Magnification Power 4X, 10X, 40XR
Model ACCU-SCOPE
Objective Types Infinity Corrected Plan
Product Type Trinocular LED Microscope
Voltage 110/240 V