Description
- Trinocular viewing head inclined at 30° and rotatable 360° for flexible viewing and camera integration.
- WF10X/20 mm focusing eyepieces with eyeguards for comfortable and clear observation.
- Reversed quintuple nosepiece with infinity-corrected PLAN 4X, 10X, 40XR, and 100XR oil objectives for precise imaging.
- Mechanical stage with low-position coaxial controls and right-hand operation for smooth slide handling.
- N.A.1.25 Abbe condenser with iris diaphragm for improved light control and contrast.
- Coaxial coarse and fine focusing controls for accurate and stable focusing adjustments.
- 3 watt variable LED illumination with field iris and dust cover for consistent lighting and protection.
Specifications
Specifications
| Type | Objective |
| Color | White |
| Voltage | 110/240 V |