Learn More
ACCU-SCOPE Trinocular Microscope with Turret Phase and Darkfield System
SDP
Catalog No. 01189341 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Magnification Power:
10X, 40X, 100X
Catalog No. Magnification Power
01-189-341 10X, 40X, 100X
1 options
Catalog No. 01-189-341 Supplier ACCU-SCOPE Supplier No. EXC350PHDF
Add to Cart
Add to Cart
Brightfield and darkfield microscope delivers high optical performance and versatility for clinical, veterinary, and educational labs.
  • Trinocular viewing head inclined 30° rotatable 360°
  • WF10X20 mm focusing eyepieces with eyeguards
  • Reversed quintuple nosepiece
  • NA 1.25 turret phase condenser 10X 40X and 100X phase annuli and objectives No 20X PH annuli or objective BF and DF positions
  • Mechanical stage with low position coaxial controls right hand control
  • Coaxial coarse and fine focusing controls 3 watt variable LED with field iris and dust cover
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Trinocular, 30° Inclined, 360° Rotating
Lighting Type LED, Darkfield
Magnification Power 10X, 40X, 100X
Model ACCU-SCOPE
Objective Types Plan Phase
Product Type Trinocular LED Microscope
Voltage 110/240 V