Learn More
ACCU-SCOPE Trinocular Microscope with Turret Phase System
SDP
Catalog No. 01189356 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Trinocular
Catalog No. Head Style
01-189-356 Trinocular
1 options
Catalog No. 01-189-356 Supplier ACCU-SCOPE Supplier No. EXC350PH
Add to Cart
Add to Cart
Provides high optical performance for labs with advanced phase contrast for bright, sharp imaging and versatile microscope applications.
  • Trinocular viewing head inclined 30° rotatable 360°
  • WF10X20 mm focusing eyepieces with eyeguards
  • Reversed quintuple nosepiece
  • NA 1.25 turret phase condenser 10X 20X 40X and 100X phase annuli and objectives and centering telescope
  • Mechanical stage with low position coaxial controls right hand control
  • Coaxial coarse and fine focusing controls
  • 3 watt variable LED with field iris and dust cover
Specifications
Eyepiece WF 10X/20 mm, with rubber eyeguard
Head Style Trinocular
Lighting Type LED
Magnification Power 10X, 20X, 40X, 100X
Model ACCU-SCOPE
Objective Types Plan Achromatic Phase
Product Type Trinocular LED Microscope
Voltage 100/240 V