Description
- Professional trinocular stereo microscope with a 6:1 zoom ratio
- Continuous magnification from 7.5X to 45X
- Integrated trinocular port for digital imaging and camera attachment
- 110 mm working distance supports larger specimen viewing
- Top and bottom LED illumination with selectable operating modes
- Designed for industrial, educational, and documentation applications
Specifications
Specifications
| Voltage | 110/220 V |
| Magnification Power | 0.75X to 4.5X |
| Objective Types | Achromatic |
| Product Type | Stereo Microscope |
| Head Style | Trinocular |
| Lighting Type | LED |
| Eyepiece | WF 10X |