Learn More
World Precision Instrument Trio 3-in-1 Analgesia System
SDP
Catalog No. 502538777
Encompass
Get three test systems in one package with the Trio, featuring the Electronic von Frey, Plethysmometer and Randall Selitto meters. Just like the Quattro package, the modular design allows these three test systems to communicate with the same electronic controller. The stand and sling for the Randall Selitto test are sold separately.
Three test systems
Catalog No. 50-253-8777 Supplier World Precision Instrument Supplier No. II2888
Add to Cart
Add to Cart
Request a Quote
Get three test systems in one package with the Trio, featuring the Electronic von Frey, Plethysmometer and Randall Selitto meters. Just like the Quattro package, the modular design allows these three test systems to communicate with the same electronic controller. The stand and sling for the Randall Selitto test are sold separately.
Three test systems