Learn More
World Precision Instrument Trio MPC100 Controller, 1 manipulators, Stainless
SDP
Catalog No. 502538533
Encompass
This motorized micromanipulator controls one or two manipulators with sub-micron (less than  100 mm) resolution. It offers a user-selectable angle from 0 - 90 degrees via a ROE input. With Fast movement with top speed of 3mm/sec (while homing), the MCP100 Manipulator Controller (WPI #: 505091) speeds up your workflow. It is compact, fanless, and user friendly. This ROE controller (WPI #: 505093) preserves bench and rack space. Multiple push button functions give you ready access to Work, Home, Lock, Pulse, Relative, Speed, and Angle. DIP switches on the ROE select the direction of movement produced by turning the ROE knob. A USB interface is available for computer control.
Catalog No. 50-253-8533 Supplier World Precision Instrument Supplier No. SUMPC145S
Add to Cart
Add to Cart
Request a Quote
This motorized micromanipulator controls one or two manipulators with sub-micron (less than  100 mm) resolution. It offers a user-selectable angle from 0 - 90 degrees via a ROE input. With Fast movement with top speed of 3mm/sec (while homing), the MCP100 Manipulator Controller (WPI #: 505091) speeds up your workflow. It is compact, fanless, and user friendly. This ROE controller (WPI #: 505093) preserves bench and rack space. Multiple push button functions give you ready access to Work, Home, Lock, Pulse, Relative, Speed, and Angle. DIP switches on the ROE select the direction of movement produced by turning the ROE knob. A USB interface is available for computer control.