Learn More
Aviva Systems Biology Corp TRIOBP Antibody - middle region (ARP39821_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39821_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIOBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIOBP. Alternate names: DFNB28, FLJ39315, HRIHFB2122, KIAA1662, TARA, dJ37E16.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57191